VWR® General Purpose Refrigerator Freezer Combination Units
Proveedor: VWR
VWR®
SCRFC-20A
97058-842EA 81685.17 MXN
97058-842
VWR® General Purpose Refrigerator Freezer Combination Units
Refrigeradores
General-purpose, upright laboratory refrigerator/freezer combination units feature a mechanical dial thermostat, leveling legs, integrated handles, interior lights, and probe access port.
  • Features heavy-duty reinforced handle and a magnetic door gasket for a tight seal
A spacious capacity allows for maximum storage space and flexibility for all types of materials. Unit operates with CFC–free refrigerant, foam insulation, and a mechanical thermostat.
Información para pedidos: Interior dimensions are subject to change without notice. Refrigerators and Freezers are supplied with a two-year parts and labor warranty, with an additional three-year warranty on compressor parts.