VWR® Four-Channel Electronic Timer and Clock with Certificate of Calibration
Proveedor: VWR
VWR®
61700-3311
89203-042CS 15952.61 MXN
89203-042
VWR® Four-Channel Electronic Timer and Clock with Certificate of Calibration
Temporizadores Timers digitales
This 4-channel timer has a capacity of 99 hours, 59 minutes and 59 seconds or can be switched to a 12- or 24-hour clock.
  • Time set memory
  • Easily portable
It counts either up or down and remembers the last time set. Lay flat on a bench, use the flip-open stand for a vertical view, or carry it on a lab coat with the convenient pocket clip.
Power supply: (1) AAA battery
Certificaciones: CE marked, RoHS compliant.
Información para pedidos: Features a Certificate of Calibration and individual serial number, these electronic timers are tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.