Boekel Scientific Digital Dry Block Heaters are designed for a variety of applications including restriction digests, C.O.D
Dry Block Heaters feature an ambient +5° to 120°C temperature range, with ±0.25°C at 37°C uniformity. Integrated into the quality design of these units is an over-temp safety feature providing safe heating of all samples.
denaturing DNA, BUN, melting agar, coagulation studies, in situ hybridization, and Hot Start PCR. The "set it and forget it" convenience along with the excellent temperature stability and rapid heat-up make this unit ideal for virtually every application. The large LED display allows for easy visibility from anywhere in the lab. These units are available in either two or four block capacity, and accommodate a variety of Dry Block Modules for added flexibility.
Certificaciones: UL listed. CE certified. CSA approved.
Información para pedidos: Boekel Scientific Digital Dry Block Heaters are supplied with a block extractor for safe removal of the blocks, instruction manual, and power cord. Each unit comes with the Boekel two-year warranty on parts and labor. Dry Block Modules sold seperately (see 35790-000 series).