Flask Shakers, Boekel Scientific
320100 320100-2 401000 401000-2
89205-644EA 79684.63 MXN
89205-644 89205-648
Flask Shakers, Boekel Scientific
The Boekel Scientific Variable and Fixed Speed Flask shakers are designed to shake up to 450 Oscillations per minute in order to simulate the back and forth motion of the wrist
The unit comes equipped with adjustable finger clamps and can accommodate up to 8 vessels or flasks. With the use of accessories, the unit can accommodate up to a total of 16 vessels or flasks. Each Flask Shaker model has a maximum load capacity of 10 pounds and for your safety and the safety of the unit, is also equipped with overload protection.
You may choose a Fixed Speed Flask Shaker which is fixed at 400 Oscillations or the Variable Speed Flask Shaker which shakes up to 450 Oscillations. Each model has an adjustable mixing amplitude of 0-15 degrees which can be set to achieve an action from a gentle swirl to a very vigorous mixing with one simple adjustment. The built in time can also be set for continuous shaking or from 1 to 60 minutes of operation.
Certificaciones: UL and CSA Certified, CE Listed
Información para pedidos: Unit comes equipped to accommodate up to 8 vessels or flasks, via two side arms and eight adjustable finger clamps.