pH-Indicator Paper Litmus Roll, MilliporeSigma
Proveedor: MilliporeSigma
1.09486.0003 1.09489.0003 1.09489.0003 1.09486.0003
EM1.09489.0003CS 237375.25 MXN
EM1.09489.0003 EM1.09486.0003 CA1.09489.0003 CA1.09486.0003
pH-Indicator Paper Litmus Roll, MilliporeSigma
Papeles de pruebas Tiras indicadoras de pH
Roll with color scale Ph Eur.
pH indicator papers supplied in rolls are better suited for longterm storage, since they are protected against external influences. pH indicator papers are manufactured by impregnating high-quality filter paper with indicator or mixed-indicator solutions. Therefore, the paper is dryed, cut and packed into the corresponding sizes.