Total Hardness Test Method: Colorimetric With Test Strips (5 Reaction Zones) MQuant™, MilliporeSigma
Proveedor: MilliporeSigma
1.10046.0001 1.10046.0001 1.10047.0013 1.10047.0013
EM1.10046.0001CS 364080 MXN
EM1.10047.0013 CA1.10047.0013 CA1.10046.0001 EM1.10046.0001
Total Hardness Test Method: Colorimetric With Test Strips (5 Reaction Zones) MQuant™, MilliporeSigma
Kits para test medioambiente
5 Reaction Zones: >5 - >10 - >15 - >20 - >25°d >6 - >13 - >19 - >25 - >31°e >9 - >18 - >27 - >36 - >45°f
MilliporeSigma # 1.10046.0001 replaces and is identical to MilliporeSigma # 10046-1
The total hardness test can be used to measure water hardness. Water hardness is caused by the presence of alkaline earths, primarily calcium and magnesium. Total hardness is measured as the sum of all calcium and magnesium ions.