RP Modified TLC Silica Gel 60 Sheets and Plates, MilliporeSigma
Proveedor: MilliporeSigma
1.15684.0001 1.13124.0001 1.15423.0001 1.14296.0001 1.05747.0001 1.05560.0001 1.05559.0001 1.15685.0001 1.15388.0001 1.15389.0001 1.13724.0001 1.15683.0001 1.15685.0001 1.05746.0001 1.13725.0001 1.13726.0001 1.14296.0001
EM1.05746.0001EA 17173.26 MXN
EM1.15683.0001 EM1.05746.0001 EM1.05559.0001 EM1.05560.0001 EM1.13124.0001 EM1.05747.0001 EM1.14296.0001 EM1.15685.0001 EM1.15388.0001 EM1.13726.0001 EM1.13724.0001 CA1.15685.0001 EM1.15389.0001 EM1.13725.0001 EM1.15423.0001 CA1.14296.0001 EM1.15684.0001
RP Modified TLC Silica Gel 60 Sheets and Plates, MilliporeSigma
Placas TLC
RP-modified plates serve two purposes: Act as a pilot method for HPLC and allow to choose various solvent system for special separations
Product # 1.05560.0001 replaces and is identical to Product # 5560-4.
Product # 1.05746.0001 replaces and is identical to Product # 5746-7.
Product # 1.05747.0001 replaces and is identical to Product # 5747-7.
Product # 1.13124.0001 replaces and is identical to Product # 13124-1.
Product # 1.13724.0001 replaces and is identical to Product # 13724-5.
Product # 1.13725.0001 replaces and is identical to Product # 13725-5.
Product # 1.13726.0001 replaces and is identical to Product # 13726-5.
Product # 1.14296.0001 replaces and is identical to Product # 14296-6.
Product # 1.15388.0001 replaces and is identical to Product # 15388-7.
Product # 1.15389.0001 replaces and is identical to Product # 15389-7.
Product # 1.15423.0001 replaces and is identical to Product # 15423-6.
Product # 1.15683.0001 replaces and is identical to Product # 15683-3.
Product # 1.15684.0001 replaces and is identical to Product # 15684-1.
Product # 1.15685.0001 replaces and is identical to Product # 15685-1.
RP-modified silica layers are well suited for many separation challenges that unmodified silica cannot overcome. These layers use aqueous solvent systems to separate extremely non-polar substances and analyze particular polar substances that can adapt to ion-pair chromatography. What's more, RP-modified silica layers are less dependent on atmospheric humidity. Unlike unmodified silica, RP-phases do not exhibit catalytic activity. This makes them the plates of choice for unstable substance that tend to experience oxidative degradation.