VWR® Powder Funnels, Wide Stem
Proveedor: VWR
H14660-0100 H14660-0150 H14660-0080 H14660-0065
16126-912CS 3874.01 MXN
16126-912 16126-916 10033-458 10033-456
VWR® Powder Funnels, Wide Stem
Embudos Embudos para polvos
Polypropylene funnels are steam autoclavable at 121°C (250°F).
  • Wide stem minimizes bridging of powder
  • 60° cone angle
  • Polypropylene construction