VWR® B.O.D. Bottles without Barcode, 300 ml
Proveedor: Avantor
284.450.02 284.452.02 284.451.02
89429-198CS 7304.87 MXN
89429-198 89429-202 89429-206
VWR® B.O.D. Bottles without Barcode, 300 ml
Frascos Frascos de DBO
Bottles conform to ASTM E438, Type I, Class B, borosilicate glass.