VWR® B.O.D. Bottles, 60 ml
Proveedor: VWR
284.242.01 284.243.01 284.241.01 284.453.01 284.450.01 284.240.01
89429-196CS 8224.2 MXN
89429-196 89429-194 89429-188 89429-192 89429-208 89429-190
VWR® B.O.D. Bottles, 60 ml
Frascos Frascos de DBO
Made from borosilicate glass, these bottles can be used for incubating diluted samples of polluted water, industrial wastes and more.
They come with a permanently screened label and are available with glass stoppers.