VWR® Narrow Mouth Reagent Bottles with Glass Stoppers
Proveedor: Avantor
272.207.04 272.207.05 272.207.07 272.207.03A 272.207.06
89429-178CS 1704.5 MXN
89429-184 89429-182 89429-178 89429-186 89429-180
VWR® Narrow Mouth Reagent Bottles with Glass Stoppers
Frascos Botellas de vidrio
Made from Borosilicate glass, these reagent bottles have necks with standard taper stoppers.