Recovery Flask, Glassco
Proveedor: Glassco
069.470.10 069.470.04 069.470.07 069.470.06 069.470.20 069.470.08 069.470.18 069.470.24 069.470.22 069.470.16 069.470.02 069.470.09 069.470.12
89426-808EA 179.9 MXN
89426-808 89426-818 89426-806 89426-816 89426-804 89426-814 89426-802 89426-812 89426-800 89426-822 89426-810 89426-820 89426-798
Recovery Flask, Glassco
Matraces Matraces de fondo redondo
Manufactured from Borosilicate 3.3 glass
All products listed are hand blown from tubing to ensure uniform thickness.