Reducing Adapter, Glassco
Proveedor: Glassco
013.470.10 013.470.02A 013.470.04 013.470.13 013.470.02
89426-332EA 230.21 MXN
89426-324 89426-332 89426-330 89426-328 89426-326
Reducing Adapter, Glassco
Adaptadores Adaptadores con juntas NS
Reducing adapters are used for connecting dissimilar standard taper joints.
  • Top [ST] joint
  • Bottom [ST] joint
Reducing adapters are equipped with joints at top and bottom.