Enlarging Adapter, Glassco
Proveedor: Glassco
014.470.03 014.470.09A 014.470.05 014.470.01 014.470.02
89426-342EA 215.99 MXN
89426-334 89426-342 89426-340 89426-338 89426-336
Enlarging Adapter, Glassco
Adaptadores Adaptadores con juntas NS
Enlarging adapters are useful for attaching apparatus with one joint size to equipment having a larger joint size.
  • Top [ST] joint
  • Bottom [ST] joint
Enlarging adapters are equipped with joints at top and bottom.