Distillation Connecting Adapter, Glassco
Proveedor: Glassco
028.470.02 028.470.04 028.470.03
89426-368EA 280.02 MXN
89426-368 89426-366 89426-372
Distillation Connecting Adapter, Glassco
Componentes para destilación Adaptadores para destilación
Adapters are equipped with standard taper outer joint at the top, and inner joints on the bottom and sidearm.
  • Top [ST] outer joint
  • Bottom and sidearm inner joints
Adapter is designed for use as a component in atmospheric or vaccum distillations.