Vacuum Take-Off Adapter, Short Stem, Glassco
Proveedor: Glassco
022.471.01 022.470.05 022.470.03
89426-400EA 188.03 MXN
89426-356 89426-400 89426-398
Vacuum Take-Off Adapter, Short Stem, Glassco
Adaptadores Adaptadores con juntas NS
Vacuum take-off adapters are used to connect a condenser to a flask for vacuum distillation.
  • Hose connection
  • Bottom drip tip
  • Short stem
Adapter features a hose connection and a drip tip at the bottom.