West Condenser, Glassco
Proveedor: Glassco
182.470.02B 182.470.01 182.470.00 182.470.03 182.470.06 182.470.02A 182.470.02
89426-658EA 339.48 MXN
89426-654 89426-664 89426-660 89426-670 89426-658 89426-668 89426-666
West Condenser, Glassco
Condensadores
Condensers feature narrow annular space for fast coolant speed, thus ensuring high cooling efficiency.