Filtration Apparatus, 47mm Fritted, 40/35, Complete Assembly, Glassco
Proveedor: Glassco
258.202.01
89426-768EA 4948.85 MXN
89426-768
Filtration Apparatus, 47mm Fritted, 40/35, Complete Assembly, Glassco
Embudos Embudos de filtración Sistema de filtros de vidrio
The unit is recommended for general filtration
Applications include, filtering corrosive solutions, particulate removal from HPLC solvents, and filtration analysis with solid phase extraction disks. Support base has a sealed-in, coarse porosity fritted disc.