Non-Welded Telescoping Base Stands, Labconco
Proveedor: Labconco
3401004 3730500 3401006 3401005 3401003
89203-038EA 11430.52 MXN
89203-038 89422-856 89422-854 89422-852 89422-850
Non-Welded Telescoping Base Stands, Labconco
Soportes de retortas
Non-Welded Telescoping Base Stands have telescoping legs that may be adjusted to eight height positions at installation in 1" increments to provide a working height from 30" to 37".
  • Easily assembled with simple hand tools
  • Casters replace the fixed feet included with Telescoping Base Stands
  • Installation is required
Non-welded stands are comprised of a durable 1.75" tubular, powder-coated steel frame with back panel support
Caster Kit includes four 5" diameter, non-marking polyurethane casters with toe locks. Load rating for each caster is 250 pounds. Casters have bearings for quiet operation. Casters add 11" (28 cm) to the overall depth of the base stand.
Four fixed, leveling fee are provided. The stand includes a 9" shelf, supports loads up to 1000 pounds. Fits an appropriately sized Purifier Logic+ Biosafety Cabinet, Purifier Cell Logic+ Biosafety Cabinet or PuriCare Procedure Station.
Certificaciones: ADA-compliant.
Información para pedidos: Caster Kit is available and sold separately. Not for use with stands that support Purifier Logic Class II, Type B2 Safety Cabinets.