Cellulose Sheets And Plates, MilliporeSigma
Proveedor: MilliporeSigma
1.05716.0001 1.05565.0001 1.05552.0001 1.05718.0001 1.15035.0001 1.05632.0001 1.05787.0001 1.05730.0001 1.05786.0001 1.05574.0001 1.15036.0001 1.16092.0001 1.05577.0001 1.05728.0001
EM1.05716.0001EA 11870.73 MXN
EM1.05718.0001 EM1.05632.0001 EM1.16092.0001 EM1.05577.0001 EM1.05728.0001 EM1.15035.0001 EM1.05574.0001 EM1.05716.0001 EM1.05565.0001 EM1.05787.0001 EM1.05552.0001 EM1.15036.0001 EM1.05730.0001 EM1.05786.0001
Cellulose Sheets And Plates, MilliporeSigma
Placas TLC
Cellulose plates are used to analyze polar substances
Product # 1.05552.0001 replaces and is identical to Product # 5551-7.
Product # 1.05574.0001 replaces and is identical to Product # 5574-7.
Product # 1.05577.0001 replaces and is identical to Product # 5577-7.
Product # 1.05632.0001 replaces and is identical to Product # 5632-5.
Product # 1.05716.0001 replaces and is identical to Product # 5716-7.
Product # 1.05718.0001 replaces and is identical to Product # 5718-7.
Product # 1.05728.0001 replaces and is identical to Product # 5728-6.
Product # 1.05730.0001 replaces and is identical to Product # 5730-6.
Product # 1.05786.0001 replaces and is identical to Product # 5786-6.
Product # 1.05787.0001 replaces and is identical to Product # 5787-5.
Product # 1.15035.0001 replaces and is identical to Product # 15035-5.
Product # 1.15036.0001 replaces and is identical to Product # 15036-6.
Product # 1.16092.0001 replaces and is identical to Product # 16092-1.
An organic sorbent, cellulose is perfect for separating hydrophilic substances by partition chromatography. Typical applications include the analysis of amino acids, carbohydrates, and phosphates as well as nucleic acid and nucleic acid derivatives. MilliporeSigma offers cellulose plates in two grades: TLC for conventional and HPTLC for demanding, high-performance separations.