Thin Layer Plates And Sheets, LiChrospher® Silica Gel 60, MilliporeSigma
Proveedor: MilliporeSigma
1.15445.0001 1.05646.0001 1.05646.0001 1.05586.0001 1.05647.0001 1.05647.0001
EM1.05586.0001EA 22036.68 MXN
EM1.15445.0001 EM1.05646.0001 CA1.05647.0001 EM1.05647.0001 CA105646.0001 EM1.05586.0001
Thin Layer Plates And Sheets, LiChrospher® Silica Gel 60, MilliporeSigma
Placas TLC
Unique HPTLC Lichrospher® plates are the first thin layer chromatography plates based on spherical silica particles
Product # 1.05586.0001 replaces and is identical to Product # 105586-1.
Product # 1.15445.0001 replaces and is identical to Product # 115445-1.
They offer the ultimate in thin layer chromatography performance and speed enabling high throughput analysis of complex samples.