VWR® Black Conductive Bags
Proveedor: VWR, part of Avantor
VWR®
6560
89072-274CS 3929.91 MXN
89072-274
VWR® Black Conductive Bags
Bolsas Bolsas para muestras
Constructed of heavy-duty 4 mm black carbon-impregnated low density polyethylene (LDPE) film to create an extra strong static barrier.
  • Polyethylene fabrication
  • Can be heat sealed, tied, stapled, or taped shut
Protects against electrostatic charge, chemicals, abrasion and humidity if heat-sealed.
Certificaciones: Meets specifications of MIL-P-82646, Rev. B.