Cryovial® Workstation Rack for Cryogenic Vials, Simport Scientific
Proveedor: Simport
T315-10 T315-2 T315-2 T315-3 T315-3 T315-10
89492-706CS 1387.39 MXN
CA48648-123 CA48648-121 89492-706 CA48648-120 89492-708 89492-710
Cryovial® Workstation Rack for Cryogenic Vials, Simport Scientific
Gradillas Gradillas de viales criogénicos
This handy autoclavable rack can hold up to 50 to 52 cryogenic vials.
Made of polypropylene.
Thanks to an innovative universal locking system, the vials will securely lock in each well and will not turn. Each position is identified with an alphanumeric index. Strong handles make it easy and safe to carry. It is supported by anti-skid rubber feet. The rack is compact and stackable.