HPTLC Silica Gel 60 Plates and Sheets, MilliporeSigma
Proveedor: MilliporeSigma
1.12572.0001 1.13192.0001 1.15534.0001 1.16464.0001 1.05641.0001 1.12572.0001 1.05649.0001 1.05547.0001 1.05642.0001 1.13749.0001 1.15552.0001 1.05644.0001 1.05649.0001 1.05616.0001 1.05628.0001 1.15647.0001 1.05629.0001 1.05556.0001 1.05636.0001 1.12668.0001 1.13192.0001 1.13749.0001 1.05616.0001 1.15696.0001 1.05548.0001 1.15534.0001 1.05633.0001 1.05631.0001
10143-780EA 16902.05 MXN
EM1.15647.0001 CA1.15534.0001 EM1.16464.0001 EM1.13192.0001 EM1.05629.0001 10143-780 EM1.05644.0001 EM1.05547.0001 CA1.05616.0001 EM1.05636.0001 EM1.13749.0001 EM1.15534.0001 EM1.05641.0001 EM1.05633.0001 CA1.12572.0001 EM1.05628.0001 CA1.13192.0001 EM1.05556.0001 EM1.05642.0001 EM1.05548.0001 CA1.05649.0001 EM1.15696.0001 EM1.12668.0001 EM1.05616.0001 CA1.13749.0001 EM1.05631.0001 EM1.15552.0001 EM1.12572.0001
HPTLC Silica Gel 60 Plates and Sheets, MilliporeSigma
Placas TLC
HPTLC plates deliver fast and quantitative analysis of complex samples for manual or automated use.
HPTLC silica plates work three times faster than conventional TLC plates - and they're more sensitive
MilliporeSigma # 1.05547.0001 replaces and is identical to MilliporeSigma # 5547-7.
MilliporeSigma # 1.05616.0001 replaces and is identical to MilliporeSigma # 105616-2.
MilliporeSigma # 1.05628.0001 replaces and is identical to MilliporeSigma # 5628-5.
MilliporeSigma # 1.05629.0001 replaces and is identical to MilliporeSigma # 5629-5.
MilliporeSigma # 1.05631.0001 replaces and is identical to MilliporeSigma # 5631-5.
MilliporeSigma # 1.05633.0001 replaces and is identical to MilliporeSigma # 5633-5.
MilliporeSigma # 1.05641.0001 replaces and is identical to MilliporeSigma # 5641-6.
MilliporeSigma # 1.05642.0001 replaces and is identical to MilliporeSigma # 5642-6.
MilliporeSigma # 1.05644.0001 replaces and is identical to MilliporeSigma # 5644-5.
MilliporeSigma # 1.13749.0001 replaces and is identical to MilliporeSigma # 13749-6.
MilliporeSigma # 1.15552.0001 replaces and is identical to MilliporeSigma # 15552-6.
MilliporeSigma # 1.15696.0001 replaces and is identical to MilliporeSigma # 15696-6.
The particle sizes range in HPTLC is between 4-8 µm, which creates a smoother surface and a higher separation power than TLC plates.