Multiformat Pre-Scored Plates, Silica Gel 60 F254, MilliporeSigma
Proveedor: MilliporeSigma
1.05620.0001 1.05608.0001 1.05620.0001 1.05635.0001
EM1.05608.0001EA 7481.48 MXN
EM1.05620.0001 EM1.05635.0001 EM1.05608.0001 CA1.05620.0001
Multiformat Pre-Scored Plates, Silica Gel 60 F254, MilliporeSigma
Placas TLC
These pre-scored glass plates fold into smaller formats with just one easy snap of the fingers.
Product # 1.05608.0001 replaces and is identical to Product # 5608-7.
Product # 1.05620.0001 replaces and is identical to Product # 105620-7.
Product # 1.05635.0001 replaces and is identical to Product # 5635-5.
Envase: Packaged in cardboard box.