Pressure Equalizing Funnels, Graduated with PTFE Needle Valve, Ace Glass Incorporated
Proveedor: Ace Glass
7298-05 7298-24 7298-28 7298-20 7298-10 7298-15
89058-754EA 12709.05 MXN
89058-748 89058-750 89058-752 89058-754 89058-756 89058-758
Pressure Equalizing Funnels, Graduated with PTFE Needle Valve, Ace Glass Incorporated
Embudos Embudos de separación
Threaded stopcock with PTFE plug permits smooth needle valve adjustment down to 0.1 mL/min
flow rate. Double PTFE ring seals prevent exposure of backup o-ring to corrosive liquids. Angled position makes manipulation of stopcock easier than conventional style. Standard Taper 24/40 joints top and bottom, double scale graduations. Fabricated from borosilicate glass.
Información para pedidos: For Replacement stopcocks, For Size 0-3 mm (89062-434) For Size 0-5 mm (89062-436).