Saturated KCl Filling Solution, VWR®
Proveedor: VWR International
89236-508 89236-512 89236-510
89236-508EA 304.52 MXN
89236-508 89236-512 89236-510
Saturated KCl Filling Solution, VWR®
Electrolitos Electrolitos de relleno
For refilling appropriate sympHony® electrodes.
  • Compatible with waterproof electrodes
  • Easy to dispense
  • Provided in a variety of volumes
For all calomel single-junction electrodes, outer chamber of Ag/AgCl double-junction electrodes, fluoride combination and sodium combination ISEs, and outer chamber of accupHast and accuTupH electrodes.