pH Electrode Storage Solution, VWR®
Proveedor: Avantor
89236-516
89236-516EA 954.07 MXN
89236-516
pH Electrode Storage Solution, VWR®
Electrolitos Soluciones de almacenamiento para electrodos
Storage solution for sympHony® pH electrodes.
  • Storage solution is pre-mixed for ready use
  • Convenient bottle size
  • Allows easy maintenance of pH electrodes
Proper pH electrode storage will maximize electrode performance and extend electrode life.