Filling Solution for Dissolved Oxygen Probe, VWR®
Proveedor: VWR International
89236-518
89236-518EA 915.86 MXN
89236-518
Filling Solution for Dissolved Oxygen Probe, VWR®
Electrolitos Electrolitos de relleno
Electrolyte filling solution for sympHony® dissolved oxygen probes.
  • Good electrolyte flow
  • Good response time of electrode
  • Easy to dispense for refills
Electrolyte filling solution is for use in double junction and ion-selective electrodes.