VWR® Conductivity Standard (22.0 ppm as NaCl)
Proveedor: VWR Chemicals BDH®
VWR®
89236-540 89236-546
89236-540EA 678.04 MXN
89236-540 89236-546
VWR® Conductivity Standard (22.0 ppm as NaCl)
Normativas Estándar para conductividad
NaCl Standard Solution is offered to calibrate instruments that measure any sample that is predominately NaCl (sodium chloride), such as seawater, brackish water, etc.
  • Sodium chloride standard solution
  • For calibration and accuracy checks in conductivity measurements
  • 500 ml bottle
Sodium chloride conductivity standard is used for calibration and accuracy checks in conductivity measurements.