VWR® Singlet™ Single-Use pH Buffer Pouches (NIST)
Proveedor: Avantor
89236-550 89236-548 89236-552
89236-552EA 68.3 MXN
89236-552 89236-548 89236-550
VWR® Singlet™ Single-Use pH Buffer Pouches (NIST)
Tampones Tampones pH
For sympHony® pH meter standardization.
  • Ideal for on-site analyses
  • Easy to use
  • Fresh and reliable
Sealed foil pouches of pH standardization buffers.