Stablcal® Turbidity Standards Calibration Kit, 2100P Portable Turbidimeter, Sealed Vials, Hach
Proveedor: Hach
Stablcal®
2659410 2659410 2659405 2659405
97009-528EA 12500 MXN
97009-528 CA26594-05 97009-550 CA97009-550
Stablcal® Turbidity Standards Calibration Kit, 2100P Portable Turbidimeter, Sealed Vials, Hach
Normativas Estándar para turbidedad
Stablcal® Calibration Standards Kit, for 2100P Portable Turbidimeter.
  • Precise and pre-mixed formazin solutions
  • Based on formazin primary standards
  • Enhanced stability
  • Saves time and labor
  • Confidence in results
Available as sealed vial format or bottles
Información para pedidos: Kit includes <0.1, 20, 100 and 800 NTU standards.