VWR® Utility Trays, Polypropylene
Proveedor: VWR, part of Avantor
16300-6216 16300-6020 16300-6226 16300-6520 16300-6818 16300-6015
89501-654EA 6073.76 MXN
89501-654 89501-656 89501-652
VWR® Utility Trays, Polypropylene
Made from polypropylene, these straight-sided trays offer resistance to temperatures up to 135°C (274°F).
  • Autoclavable
  • Polypropylene fabrication