VWR® Standard Standard Spectrophotometer Cuvettes
Proveedor: VWR
VWR®
K1960-VWR K1961-VWR
58017-875CS 3163.13 MXN
58017-880 58017-875
VWR® Standard Standard Spectrophotometer Cuvettes
Cubetas Cubetas para espectrofotómetros
VWR® Standard Cuvettes feature four optical windows that are recessed to prevent scratching.
  • Recessed windows
  • Polystyrene or acrylic construction
  • Suitable for accurate UV visible analyses
Spectrophotometer cuvettes are manufactured in a variety of clear materials to support different wavelength range and pathlength requirements.