VWR® Basic Chromatography Refrigerators with Glass Doors and Natural Refrigerant
Proveedor: VWR
VWR®
GDM49WHTSSPDM GDM72WHTSSPDM GDM47HC2P1D1M GDM23WHTSSPDM GDM47HCS2P1D1M GDM23WHTSSPD GDM23HC1P1D1M GDM49HC2P1D1M GDM41HC2P1D1M GDM72HC2P1D1M GDM41HCS2P1D1M GDM23HC1P1D
10018-858EA 134051.32 MXN
10018-856 10018-868 10018-858 75840-438 10791-614 10791-616 10018-862 10791-628 75794-804 75840-440 10791-622 75794-802
VWR® Basic Chromatography Refrigerators with Glass Doors and Natural Refrigerant
Refrigeradores
Swing door, upright laboratory chromatography refrigerators maintain an average temperature of 3–5°C (37–41°F).
  • Units maintain an average temperature of 3–5°C (37–41°F)
  • Equipped with energy efficient thermal insulated glass doors
  • Three-year parts and labor warranty
  • Additional two-year warranty on compressor parts
Units are equipped with low E, double-pane thermal insulated glass doors for energy efficiency
All units feature a natural refrigerant that is environmentally friendly. The R290 hydrocarbon refrigerant emits zero (0) ozone depletion potential (ODP), and three (3) global warming potential (GWP).
Units are supplied standard with the LAE temperature control, which offers exterior digital display and alarm function.
The 23 cu.ft. units equipped with half shelves feature one access port, one duplex outlet, and one chromatography mast. The 23 cu.ft. units equipped with full shelves feature one access port and one duplex outlet. The 41, 47, 49 and 72 cu.ft. units feature two access ports, one duplex outlet, and one chromatography mast.
All refrigerators are supplied standard with adjustable, heavy-duty, PVC-coated white wire shelves and LED lighting.
Certificaciones: C-UL US listed. NSF certified. Natural Refrigerant.
Información para pedidos: Additional full and half white wire shelves can be ordered seperately for any VWR® Single Glass Door Chromatography Refrigerators. Models with swing doors are available on request, please contact your local Avantor sales office for details. Optional Chart Recorder, Alarm, and Thermometer accessories require factory installation. Refrigerators come with a three-year parts and labor warranty, with an additional two-year warranty on the compressors.