NORM-JECT® Luer Slip Bulk Non Sterile Syringes, Air-Tite
Proveedor: Air-Tite
NORM-JECT®
4010.290D0 4050.090DZ 4100.090D0 4020.090D0
53547-006CS 15264.8 MXN
53547-000 53547-002 53547-004 53547-006
NORM-JECT® Luer Slip Bulk Non Sterile Syringes, Air-Tite
Jeringas Jeringas de uso múltiple
These NORM-JECT® Luer slip bulk non sterile syringes provide great value when purchased in bulk quantites.
  • These are latex free, rubber free, silicone free, styrene free and DEHP free
NORM-JECT® syringes are the choice for any situation needing an inert, non-reactive syringe. Because of their composition they are indicated for nuclear medicine, amniocentisis, IVF, embryo-transfer, chromatography and many laboratory procedures. They are more chemically resistant than rubber tipped syringes. Manufactured only from laboratory grade polypropylene and polyethylene. These unique plastic syringes have a positive safety stop to prevent accidental spills.
Certificaciones: Meets ISO standards, HPLC certified and DNA-free certified.
Envase: Bulk, non-sterile.