Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
Login
/
Register
Mexico
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
$0.00
Order Entry
Polysulfone Filter Holders, 47 mm, Advantec
101105-240
:
ADVANTEC MFS MS
Polysulfone Filter Holders, 47 mm, Advantec
101105-240
:
ADVANTEC MFS MS
:
43301020
Capacity:
300 ml
Description:
Polysulfone Aseptic Unit, Model KP-47U with Funnel, Base and Cover
Filtration area:
13.5 cm
2
Cat. no.:
101105-240
Supplier No.:
501030