VWR® General Purpose Water Baths
WB02V11B WB20V11B TMPVWRPS10L TMPVWRPS20L TMPVWRPS02L WB05V11B TMPVWRPS05L TMPVWRPS28L WB28V11B WB10V11B
89501-472EA 38901.89 MXN
89501-472 89501-460 89501-468 89501-476 10796-926 89501-464
VWR® General Purpose Water Baths
The versatile, general-purpose water bath features a digital display.
  • Two-Year Warranty
  • High-Clearance Lid
  • Three User-Configured Temperature Presets
  • Built-In Countdown Timer
  • Removable Bottom Diffuser Tray
Other features include an audible alarm, a programmable timer, high limit, and offset with three preset temperatures, also programmable.
A built-in countdown timer and manufacturer's warranty are also included.
Certificaciones: 120V units are ETL listed (North America only). ISO 17025 compliant.
Información para pedidos: This water bath includes a lid and removable bottom diffuser tray. 240V/50Hz models are also available; contact your VWR sales representative for more information.