Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Proveedor: Thermo Scientific
S7515-3 S7515-20 S7515-30 S7515-5 S7515-10
66064-770CS 30166.75 MXN
66064-770 66064-772 75801-380 66064-766 66064-768
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Jeringas Jeringas de uso múltiple
All-plastic syringes are free of rubber components or lubricants
These syringes are preferred for use with Target™ brand syringe filters. Polyethylene barrels and polypropylene plungers offer wide solvent compatibility, low background extractables and the convenience of a disposable product. No silicone or oil lubricant is required in the barrel.