VECTASHIELD® Mounting Medium, Vector Laboratories
Proveedor: Vector Laboratories
VECTASHIELD®
H-1000-10 H-1400-10 H-1300-10 H-1500-10 H-1200-10
101098-042EA 7224.02 MXN
101098-048 101098-046 101098-044 101098-042 101098-050
VECTASHIELD® Mounting Medium, Vector Laboratories
Tintes Reactivos de microscopía y tinción
VECTASHIELD® Mounting Media minimize photobleaching during fluorescence-based imaging. There are three versions: original VECTASHIELD, a non-curing formulation that allows immediate slide viewing; VECTASHIELD HardSet, which cures for long-term storage without sealing; and VECTASHIELD Vibrance™, a new hardening formulation that offers superior handling and greater stability for long-term storage of slides at room temperature.
VECTASHIELD® Mounting Media are compatible with fluorescent nuclear stains, fluorescent proteins, fluorescent tracers, histochemical stains, and most fluorochromes, with VECTASHIELD Vibrance showing superior performance with far-red dyes.
All three VECTASHIELD formulations are available with and without DAPI, a common fluorescent nuclear counterstain. DAPI excites at about 360 nm and emits at about 460 nm when bound to DNA, producing nuclei with blue fluorescence. VECTASHIELD HardSet is also available with phalloidin (PI), a bicyclic heptapeptide that stains actin filaments. PI excites at about 535 nm and emits at about 615 nm, producing a red fluorescence.
Certificaciones: Manufactured under ISO9001:2015