VWR® Wire Petri Dish Holder
HS23455
82008-018EA 682.21 MXN
82008-018
VWR® Wire Petri Dish Holder
Designed to keep dishes on hand during an experiment, the stand holds up to six 100 mm Petri dishes.
  • Epoxy-coated steel construction
  • Keep dishes on hand during experimentation
  • Suited for small-batch incubation
  • Autoclavable
Constructed of epoxy-coated steel wire, it is perfectly suited for small-batch incubation.