VWR® 80-Well Microtube Racks
Proveedor: VWR
VWR®
HS29025DV HS29025AV HS29025EV HS29025GV HS29025FV HS29025BV HS29025CV
82024-469CS 18795.47 MXN
82024-469 82024-467 82024-479 82024-473 82024-471 82024-477 82024-475
VWR® 80-Well Microtube Racks
Gradillas Tube Racks
Popular fraction-collector-plate style racks can easily fill and transfer 80 tubes in compact arrangement (5x16 array).
  • Easy organization and identification
  • Transfer up to 80 tubes in a compact arrangement
These polypropylene racks hold 1.5–2.0 mL microtubes
Assorted color coding orders include one each of Blue, Green, Orange, Pink, and Yellow.
Molded-in coordinates aid organization and identification.