Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Proveedor: Thermo Scientific
900020 910001 9100CB 900024 900023 900021-WA 911125 910060 911110 900022-WA 810001
34104-300EA 2685 MXN
34175-516 34104-300 34175-524 34104-298 34104-302 10781-884 34108-041 23198-300 34104-296 14004-272 34104-294
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Electrolitos Soluciones de almacenamiento para electrodos
Cleaning Solutions restore proper electrode performance and prolong the useful life of the electrode.
  • Restore electrode performance
  • Prolong usefulness
  • Unique formulations
Cleaning Solutions are identified by the letters A, B, C, and D. Each letter designates a unique formulation. Cleaning solution A is for removing protein contaminants. Cleaning solution B is for removing bacterial contaminants. Cleaning solution C is for general cleaning. Cleaning solution D is for removing oil and grease contaminants. Bottles include a small beaker to hold the solution and a plastic pipet to remove the old electrode filling solution.