SP Bel-Art H-B DURAC® Plus™ Individually Calibrated Liquid-In-Glass Thermometers, Organic Liquid Fill, Bel-Art Products, a part of SP
Proveedor: Bel-Art Products, a part of SP
B60205-0500 B60205-0900 B60205-0200 B60205-0800
30617-774EA 16433.42 MXN
30617-774 30617-806 30617-810 30617-778
SP Bel-Art H-B DURAC® Plus™ Individually Calibrated Liquid-In-Glass Thermometers, Organic Liquid Fill, Bel-Art Products, a part of SP
Termómetros Analogue Thermometers
Individually calibrated, these thermometers feature blue organic liquid fill against lead-free white back glass
  • Ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical/medical industry, manufacturing, dairies, breweries and distilleries
  • Individually serialized
Includes individual Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration.