Multi-Paddle Agitator, PTFE, Ace Glass Incorporated
Proveedor: Ace Glass
8089-18 8089-04 8089-14 8089-16 8089-06 8089-08
89062-124EA 4084.94 MXN
89062-124 89062-126 89062-128 89062-130 89062-122 89062-132
Multi-Paddle Agitator, PTFE, Ace Glass Incorporated
Imánes de agitación Ejes de agitación
The PTFE multi-paddle agitator has paddles that are replaceable
Fits 10 mm O.D. drilled or certain knob type shafts.