Valor® 7000 Compact Bench Scales, Ohaus
Proveedor: Ohaus
Valor®
30031827 30031829 30031830 30031831 30031828
89497-054EA 20675.16 MXN
89497-050 89497-048 89497-046 89497-054 89497-052
Valor® 7000 Compact Bench Scales, Ohaus
Balanzas
The Valor® 7000 compact food scale is built with features designed to improve food weighing and proportioning productivity
Touchless sensors located in the front and rear allow users to perform weighing without ever touching the scale. Determine weight and perform zero, tare, print, and function operations with one swift hand motion. This helps to keep work environments clean, reduce the chances of cross-contamination, and ultimately provide years of reliable service. Designed to be one of the most user-friendly products on the market, each unit features two large displays allow two operators to share one scale. Displays are easy to read even from a distance. The 1.5” front display and 1” rear display are both equipped with an LCD backlight for added clarity. Scales also feature checklights for another level of clarification and confirmation of results.
Dual displays and touchless sensors, user-friendly menu and setup, along with the speed and accuracy of the results provide for a seamless and hassle-free user experience that is second to none.
Certificaciones: Food Safety Certification (NSF) and supports a HACCP compliant quality system. This product conforms to the EMC Directive 2004/108/EC, and the Low Voltage Directive 2006/95/EC.
Información para pedidos: Ethernet, USB connectivity, and additional RS232 ports are all available as accessory items.