Automatic High-Pressure Steam Sterilizer, Yamato Scientific
SM-300 SM511 SM-200 SM-310
27585-042EA 303454.3 MXN
27585-040 27585-042 27585-034 27585-038
Automatic High-Pressure Steam Sterilizer, Yamato Scientific
The Automatic High-Pressure Steam Sterilizer features an interactive key input system, which allows users to sterilize and dry instruments or clothes used in research or examination of medicine, medical treatment, or food with the utmost safety.
  • High-pressure
  • Interactive key input system
  • Automatic processes
  • Self-diagnosis function
The interactive key input system enables conditions to be memorized
Units offer a sterilization temperature setting range of 105 to 128°C, and a drying temperature setting range of 150 to 180°C. Maximum operation pressure is 0.2MPa. Interior chamber is constructed of stainless steel.
With a single key press, everything is done automatically from air exhausting to sterilization and drying. Preset or continuous operation modes are available. Safety measures are enriched, including a self-diagnosis function and a prevention device for misplacement of drain-bottles.