Orion™ Conductivity Standards, Solutions, and Accessories, Thermo Scientific
Proveedor: Thermo Scientific
Orion™
011006 011007 011005 011005 011008 990106
52458-147CS 4254.37 MXN
52458-147 CA011005 52458-149 71002-397 52458-145 23227-504
Orion™ Conductivity Standards, Solutions, and Accessories, Thermo Scientific
Normativas Estándar para conductividad
Perform virtually any electrochemistry analysis with Orion™ Conductivity Standards and Solutions
  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
Conductivity standard temperature tables are programmed into Orion™ meter software for use with the automatic calibration feature.
Choose from a complete selection of conductivity accessories including conductivity standards and TDS (total dissolved solids) standards. All conductivity standards are traceable to NIST standard reference materials.