Orion™ DO Probe for Laboratory or Field, Thermo Scientific
Proveedor: Thermo Scientific
Orion™
083005MD 083010MD 083060MD 083005MD 083025MD
10010-710EA 21057.13 MXN
CA10010-712 10010-710 10010-716 10010-714 CA10010-710
Orion™ DO Probe for Laboratory or Field, Thermo Scientific
Electrodos Electrodos de oxígeno disuelto
Use this dissolved oxygen probe as the name implies, either in the laboratory or filed.
  • Less than 1% drift per day
  • Maximum overpressure is 6 bar
  • Minimum sample flow of 10 cm/second
  • Oxygen consumption is 0.008 µg/h (mg/L) at 20°C
  • 90% of final value in 10 seconds
  • Dual, thermally separated sensors
The Orion™ DO Probe for Laboratory or Field is durable and accurate, featuring automatic temperature compensation and convenient screw-on membrane caps.