Stomacher® 400 Lab Blender Series, Seward
Proveedor: Seward
030010108 030010159
58976-916EA 201159.84 MXN
58976-960 58976-916
Stomacher® 400 Lab Blender Series, Seward
Homogenizadores Homogenizadores de palas
Whatever method you use to enumerate and identify bacteria, yeasts and moulds in your laboratory, thorough reproducible homogenisation of the sample is the first and most critical step
Numerous studies have demonstrated the Circulator enhancement as a positive benefit to pathogen testing procedures in traditional microbiology and PCR based techniques. The Stomacher® 400 is durable and reliable, and it offers improved organism recovery with the patented curved shape of the circulator paddles over older square paddle designs.
As the sample is crushed during operation, the patented curved shape of the Circulator paddles create a washing effect driving more organisms in suspension thereby providing a better homogenised sample for analysis. There is no risk of cross contamination between samples.
The Stomacher® 400 features a removable door for easy cleaning or paddle chamber, a countdown to processing time, and a programmable control panel with 'hands-free' auto-run available.
The Stomacher® 400 meets the food safety and quality demands of today’s microbiologists.
Información para pedidos: Machine comes with 5 year warranty and 10 year service support.